Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ILR1
Entrez Gene ID
821199
Gene Name
Peptidase M20/M25/M40 family protein
Gene Aliases
AT3G02875, F13E7.18, F13E7_18, IAA-AMINO ACID HYDROLASE, IAA-LEUCINE RESISTANT 1
Chromosome Location
Chr.3: 631824 - 634074 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337440.1 NP_001325526.1 3 - 4 1186 141
NM_111156.3 NP_186937.1 4 - 5 1094 141
GenBank mRNA AY054640.1 - 4 - 5 1005 141
AY081499.1 - 4 - 5 925 141

View Full Product Details