Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VGT1
Entrez Gene ID
821117
Gene Name
vacuolar glucose transporter 1
Gene Aliases
AT3G03090, AtVGT1, T17B22.22, T17B22_22, vacuolar glucose transporter 1
Chromosome Location
Chr.3: 700433 - 704828 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111179.4 NP_186959.2 1 - 2 296 74
GenBank mRNA AY140054.1 - 1 - 2 185 74
BT010375.1 - 1 - 2 47 74

View Full Product Details