Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
XPO1B
Entrez Gene ID
821074
Gene Name
exportin 1B
Gene Aliases
AT3G03110, CRM1B, T17B22.20, T17B22_20, exportin 1B
Chromosome Location
Chr.3: 708698 - 717093 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111181.3 NP_566193.2 9 - 10 1126 141

View Full Product Details