Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DCL2
Entrez Gene ID
821300
Gene Name
dicer-like 2
Gene Aliases
AT3G03300, ATDCL2, DICER-LIKE 2, T17B22.28, dicer-like 2
Chromosome Location
Chr.3: 767880 - 776214 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084632.1 NP_001078101.1 11 - 12 1678 123
NM_001202869.2 NP_001189798.1 14 - 15 2041 123
NM_001337464.1 NP_001327050.1 8 - 9 1159 123
NM_111200.5 NP_566199.4 15 - 16 2283 123

View Full Product Details