Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DCL2
Entrez Gene ID
821300
Gene Name
dicer-like 2
Gene Aliases
AT3G03300, ATDCL2, DICER-LIKE 2, T17B22.28, dicer-like 2
Chromosome Location
Chr.3: 767880 - 776214 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084632.1 NP_001078101.1 2 - 3 459 150
NM_001202869.2 NP_001189798.1 5 - 6 822 150
NM_111200.5 NP_566199.4 6 - 7 1064 150

View Full Product Details