Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ATK1
- Entrez Gene ID
-
827876
- Gene Name
- kinesin 1
- Gene Aliases
-
AT4G21270, F7J7.210, F7J7_210, KATA, KATAP, KINESIN-LIKE PROTEIN IN ARABIDOPSIS THALIANA A, KINESIN-LIKE PROTEIN IN ARABIDOPSIS THALIANA A PROTEIN, KINESIN-LIKE PROTEIN IN ARABIDOPSIS THALIANA A, KINESIN-LIKE PROTEIN IN ARABIDOPSIS THALIANA A PROTEIN, kinesin 1
- Chromosome Location
-
Chr.4: 11329319 - 11334168 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_118246.2 |
NP_193859.1 |
7 - 8 |
1342 |
77 |
GenBank mRNA |
D11371.1 |
- |
7 - 8 |
1309 |
77 |
View Full Product Details