Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRXR1
Entrez Gene ID
828285
Gene Name
Peroxidase superfamily protein
Gene Aliases
AT4G21960, F1N20.3, PEROXIDASE PRXR1
Chromosome Location
Chr.4: 11646109 - 11648644 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118317.4 NP_567641.1 3 - 4 919 85
GenBank mRNA AF325015.2 - 3 - 4 631 85
AF428379.1 - 3 - 4 631 85
AY056809.1 - 3 - 4 631 85
AY058071.1 - 3 - 4 631 85
AY059810.1 - 3 - 4 626 85
AY132009.1 - 3 - 4 587 85
X98313.1 - - 636 85

View Full Product Details