Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIP1;5
Entrez Gene ID
828439
Gene Name
plasma membrane intrinsic protein 1;5
Gene Aliases
AT4G23400, F16G20.100, F16G20_100, PCR55, PIP1D, plasma membrane intrinsic protein 1;5
Chromosome Location
Chr.4: 12220641 - 12222468 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118469.4 NP_194071.1 3 - 4 919 77
GenBank mRNA AY059948.1 - 3 - 4 817 77
AY081593.1 - 3 - 4 768 77

View Full Product Details