Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CPSF30
Entrez Gene ID
839925
Gene Name
cleavage and polyadenylation specificity factor 30
Gene Aliases
ARABIDOPSIS THALIANA CLEAVAGE AND POLYADENYLATION SPECIFICITY FACTOR 30, AT1G30460, ATCPSF30, F26G16.6, F26G16_6, cleavage and polyadenylation specificity factor 30
Chromosome Location
Chr.1: 10770813 - 10775428 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332901.1 NP_001319113.1 5 - 6 1160 102
NM_102782.4 NP_174334.2 5 - 6 1168 102
GenBank mRNA EU250988.1 - 5 - 6 1142 102

View Full Product Details