Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHV
Entrez Gene ID
839928
Gene Name
Homeobox-leucine zipper family protein / lipid-binding START domain-containing protein
Gene Aliases
AT1G30490, ATHB9, F26G16.11, F26G16_11, PHAVOLUTA
Chromosome Location
Chr.1: 10796124 - 10801093 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102785.5 NP_174337.1 13 - 14 2024 82
GenBank mRNA AJ440967.1 - 13 - 14 1675 82
AK226570.1 - 13 - 14 1889 82

View Full Product Details