Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AAE14
Entrez Gene ID
839932
Gene Name
acyl-activating enzyme 14
Gene Aliases
AT1G30520, F26G16.14, F26G16_14, acyl-activating enzyme 14
Chromosome Location
Chr.1: 10810397 - 10813779 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332903.1 NP_001322322.1 3 - 4 1087 89
NM_001332904.1 NP_001322320.1 3 - 4 607 89
NM_001332905.1 NP_001322321.1 3 - 4 607 89
NM_102789.5 NP_174340.2 3 - 4 1087 89
GenBank mRNA AY070739.1 - 3 - 4 500 89
AY250843.1 - 3 - 4 445 89
BT010462.1 - 3 - 4 445 89

View Full Product Details