Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- OSB4
- Entrez Gene ID
-
839987
- Gene Name
- organellar single-stranded DNA binding protein 4
- Gene Aliases
-
AT1G31010, F17F8.7, organellar single-stranded DNA binding protein 4
- Chromosome Location
-
Chr.1: 11054731 - 11056959 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_102839.5 |
NP_564370.1 |
3 - 4 |
820 |
151 |
View Full Product Details