Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KUP10
Entrez Gene ID
839997
Gene Name
K+ uptake permease 10
Gene Aliases
AT1G31120, F28K20.5, F28K20_5, K+ uptake permease 10
Chromosome Location
Chr.1: 11104237 - 11108095 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102850.4 NP_174397.2 2 - 3 1110 112
GenBank mRNA AK229208.1 - 2 - 3 779 112

View Full Product Details