Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SRX
Entrez Gene ID
840004
Gene Name
sulfiredoxin
Gene Aliases
AT1G31170, ATSRX, F28K20.12, F28K20_12, SULFIREDOXIN, sulfiredoxin
Chromosome Location
Chr.1: 11133410 - 11135189 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084168.1 NP_001077637.1 4 - 5 399 107
NM_001084169.1 NP_001077638.1 4 - 5 393 107
NM_102855.4 NP_564375.1 4 - 5 424 107
GenBank mRNA AK117789.1 - 4 - 5 370 107

View Full Product Details