Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KAN2
Entrez Gene ID
840116
Gene Name
Homeodomain-like superfamily protein
Gene Aliases
AT1G32240, F27G20.7, F27G20_7, KANADI 2
Chromosome Location
Chr.1: 11625616 - 11630745 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333002.1 NP_001322279.1 3 - 4 298 145
NM_102957.4 NP_564392.1 4 - 5 1374 145
GenBank mRNA AB493491.1 - 4 - 5 984 145
AY048689.1 - 4 - 5 1002 145
BT029211.1 - 4 - 5 984 145

View Full Product Details