Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sks9
Entrez Gene ID
829999
Gene Name
SKU5 similar 9
Gene Aliases
AT4G38420, F22I13.190, F22I13_190, SKU5 similar 9
Chromosome Location
Chr.4: 17982616 - 17985384 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342496.1 NP_001329209.1 5 - 6 1661 93
NM_120004.5 NP_195555.2 6 - 7 1661 93
GenBank mRNA AY072215.1 - 6 - 7 1503 93
AY096593.1 - 6 - 7 1437 93

View Full Product Details