Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
APK
Entrez Gene ID
815963
Gene Name
APS kinase
Gene Aliases
ADENOSINE PHOSPHOSULFATE KINASE, ADENOSINE-5'-PHOSPHOSULFATE (APS) KINASE 1, AKN1, APK1, APS KINASE 1, APS kinase, AT2G14750, ATAKN1, F26C24.11, F26C24_11
Chromosome Location
Chr.2: 6313883 - 6316047 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127039.4 NP_179082.1 2 - 3 576 82
GenBank mRNA AY054287.1 - 2 - 3 423 82
AY132010.1 - 2 - 3 331 82
U05238.1 - 2 - 3 363 82
X75782.1 - - 372 82

View Full Product Details