Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
APK
Entrez Gene ID
815963
Gene Name
APS kinase
Gene Aliases
ADENOSINE PHOSPHOSULFATE KINASE, ADENOSINE-5'-PHOSPHOSULFATE (APS) KINASE 1, AKN1, APK1, APS KINASE 1, APS kinase, AT2G14750, ATAKN1, F26C24.11, F26C24_11
Chromosome Location
Chr.2: 6313883 - 6316047 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127039.4 NP_179082.1 4 - 5 849 72
GenBank mRNA AY054287.1 - 4 - 5 696 72
AY132010.1 - 4 - 5 604 72
U05238.1 - 4 - 5 636 72
X75782.1 - - 645 72

View Full Product Details