Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LIP1
Entrez Gene ID
816012
Gene Name
lipase 1
Gene Aliases
AT2G15230, ATLIP1, F15A23.3, F15A23_3, lipase 1
Chromosome Location
Chr.2: 6612480 - 6615457 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127084.3 NP_179126.2 5 - 6 833 154
GenBank mRNA AF530477.1 - 5 - 6 674 154
AK229578.1 - 5 - 6 732 154
BT025547.1 - 5 - 6 647 154

View Full Product Details