Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARFB1A
Entrez Gene ID
816020
Gene Name
ADP-ribosylation factor B1A
Gene Aliases
ADP-ribosylation factor B1A, AT2G15310, ATARFB1A, F27O10.4, F27O10_4
Chromosome Location
Chr.2: 6653663 - 6655501 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127091.3 NP_179133.1 2 - 3 504 109
GenBank mRNA AY054470.1 - 2 - 3 375 109
AY093273.1 - 2 - 3 220 109

View Full Product Details