Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PMT1
- Entrez Gene ID
-
816109
- Gene Name
- polyol/monosaccharide transporter 1
- Gene Aliases
-
ARABIDOPSIS THALIANA POLYOL/MONOSACCHARIDE TRANSPORTER 1, AT2G16120, ATPMT1, F7H1.14, F7H1_14, polyol/monosaccharide transporter 1
- Chromosome Location
-
Chr.2: 6996727 - 6998441 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_127170.1 |
NP_179209.1 |
2 - 3 |
469 |
98 |
View Full Product Details