Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- AMS
- Entrez Gene ID
-
816194
- Gene Name
- basic helix-loop-helix (bHLH) DNA-binding superfamily protein
- Gene Aliases
-
ABORTED MICROSPORES, AT2G16910, F12A24.9, F12A24_9
- Chromosome Location
-
Chr.2: 7331721 - 7334254 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_127244.4 |
NP_179283.2 |
4 - 5 |
411 |
68 |
View Full Product Details