Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SDC
- Entrez Gene ID
-
816276
- Gene Name
- ascorbic acid mannose pathway regulator, putative (DUF295)
- Gene Aliases
-
AT2G17690, SUPPRESSOR OF DRM1 DRM2 CMT3, T17A5.18, T17A5_18
- Chromosome Location
-
Chr.2: 7682162 - 7684083 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_127323.3 |
NP_179360.1 |
3 - 3 |
1277 |
77 |
- Gene Symbol
- AT2G17695
- Entrez Gene ID
-
816277
- Gene Name
- outer envelope protein
- Gene Aliases
-
AT2G17695
- Chromosome Location
-
Chr.2: 7682781 - 7685202 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_179634.4 |
NP_849965.1 |
3 - 3 |
1268 |
77 |
View Full Product Details