Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- EDA2
- Entrez Gene ID
-
816320
- Gene Name
- Serine carboxypeptidase S28 family protein
- Gene Aliases
-
AT2G18080, T27K22.5, T27K22_5, embryo sac development arrest 2
- Chromosome Location
-
Chr.2: 7857139 - 7859983 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_127364.6 |
NP_179399.5 |
10 - 11 |
1330 |
71 |
View Full Product Details