Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCC3
Entrez Gene ID
820496
Gene Name
multidrug resistance-associated protein 3
Gene Aliases
AT3G13080, ATMRP3, ATP-binding cassette C3, MJG19.26, MJG19_26, MULTIDRUG RESISTANCE PROTEIN 3, multidrug resistance-associated protein 3
Chromosome Location
Chr.3: 4195770 - 4201734 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338031.1 NP_001327928.1 5 - 6 4307 86
NM_112147.3 NP_187915.1 5 - 6 4307 86
NM_180244.1 NP_850575.1 5 - 6 3838 86
NM_202571.1 NP_974300.1 4 - 5 3953 86

View Full Product Details