Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LAC10
Entrez Gene ID
831697
Gene Name
laccase 10
Gene Aliases
AT5G01190, F7J8.170, F7J8_170, laccase 10
Chromosome Location
Chr.5: 72292 - 74769 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120197.4 NP_195739.2 1 - 2 192 63
GenBank mRNA BT014855.1 - 1 - 2 92 63

View Full Product Details