Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ESP4
Entrez Gene ID
831597
Gene Name
HEAT repeat-containing protein
Gene Aliases
AT5G01400, ENHANCED SILENCING PHENOTYPE 4, T10O8.110, T10O8_110
Chromosome Location
Chr.5: 162522 - 171302 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342587.1 NP_001330013.1 8 - 9 2372 85
NM_001342588.1 NP_001330014.1 9 - 10 2329 85
NM_120218.2 NP_195760.1 9 - 10 2093 85

View Full Product Details