The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_120241.4 | NP_195783.3 | 5 - 6 | 1764 | 122 |
GenBank mRNA | AJ488305.1 | - | 5 - 6 | 1569 | 122 |