Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
STN8
Entrez Gene ID
831741
Gene Name
Protein kinase superfamily protein
Gene Aliases
AT5G01920, STATE TRANSITION 8, State transition 8, T20L15.190, T20L15_190
Chromosome Location
Chr.5: 359107 - 361310 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342626.1 NP_001331798.1 1 - 2 614 59
NM_120270.4 NP_195812.1 1 - 2 614 59
GenBank mRNA AK117628.1 - 1 - 2 589 59
BT006132.1 - 1 - 2 567 59

View Full Product Details