Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HTA12
Entrez Gene ID
831891
Gene Name
histone H2A 12
Gene Aliases
AT5G02560, T22P11.150, T22P11_150, histone H2A 12
Chromosome Location
Chr.5: 575225 - 577216 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120334.3 NP_195876.1 1 - 2 426 91
GenBank mRNA AY085515.1 - 1 - 2 269 91
BT025612.1 - 1 - 2 214 91

View Full Product Details