Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SDH1-2
Entrez Gene ID
816359
Gene Name
succinate dehydrogenase 1-2
Gene Aliases
AT2G18450, T30D6.4, T30D6_4, succinate dehydrogenase 1-2
Chromosome Location
Chr.2: 7997344 - 8000825 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127401.3 NP_179435.1 11 - 12 1592 74
GenBank mRNA AK119142.1 - 11 - 12 1571 74
BT005938.1 - 11 - 12 1568 74

View Full Product Details