Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OFP7
Entrez Gene ID
816365
Gene Name
ovate family protein 7
Gene Aliases
ARABIDOPSIS THALIANA OVATE FAMILY PROTEIN 7, AT2G18500, ATOFP7, F24H14.15, F24H14_15, ovate family protein 7
Chromosome Location
Chr.2: 8026957 - 8028273 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127406.3 NP_179440.1 1 - 1 1169 148
GenBank mRNA BT002903.1 - 1 - 1 1092 148

View Full Product Details