Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GGPS4
Entrez Gene ID
816379
Gene Name
geranylgeranyl pyrophosphate synthase 4
Gene Aliases
AT2G18640, F24H14.2, geranylgeranyl pyrophosphate synthase 4
Chromosome Location
Chr.2: 8083518 - 8084960 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127420.2 NP_179454.1 1 - 1 705 77
GenBank mRNA DQ446521.1 - 1 - 1 617 77

View Full Product Details