The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001335621.1 | NP_001318247.1 | 5 - 6 | 1001 | 95 |
NM_001335622.1 | NP_001324425.1 | 6 - 7 | 1353 | 95 | |
NM_127468.5 | NP_179501.1 | 6 - 7 | 1155 | 95 | |
GenBank mRNA | AJ297264.1 | - | 5 - 6 | 960 | 95 |
AK227102.1 | - | 6 - 7 | 1064 | 95 | |
AY096796.1 | - | 6 - 7 | 1061 | 95 |