Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- EER5
- Entrez Gene ID
-
816475
- Gene Name
- proteasome family protein
- Gene Aliases
-
AT2G19560, AtTHP1, ENHANCED ETHYLENE RESPONSE 5, ESSP1, F3P11.16, F3P11_16, ectopic expression of seed storage proteins 1
- Chromosome Location
-
Chr.2: 8465690 - 8469145 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_127514.4 |
NP_179546.2 |
2 - 3 |
381 |
84 |
GenBank mRNA |
AK118897.1 |
- |
2 - 3 |
352 |
84 |
View Full Product Details