Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EER5
Entrez Gene ID
816475
Gene Name
proteasome family protein
Gene Aliases
AT2G19560, AtTHP1, ENHANCED ETHYLENE RESPONSE 5, ESSP1, F3P11.16, F3P11_16, ectopic expression of seed storage proteins 1
Chromosome Location
Chr.2: 8465690 - 8469145 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335638.1 NP_001324728.1 7 - 8 577 72
NM_127514.4 NP_179546.2 9 - 10 929 72
GenBank mRNA AK118897.1 - 9 - 10 900 72

View Full Product Details