Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TET2
Entrez Gene ID
816477
Gene Name
tetraspanin2
Gene Aliases
AT2G19580, F3P11.18, F3P11_18, tetraspanin2
Chromosome Location
Chr.2: 8472134 - 8475153 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127516.4 NP_179548.1 1 - 2 319 63
GenBank mRNA AK229021.1 - 1 - 2 257 63
BT011213.1 - 1 - 2 231 63
BT012532.1 - 1 - 2 187 63

View Full Product Details