Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PSB29
Entrez Gene ID
816623
Gene Name
photosystem II reaction center PSB29 protein
Gene Aliases
AT2G20890, F5H14.14, F5H14_14, THF1, THYLAKOID FORMATION1, photosystem II reaction center PSB29 protein
Chromosome Location
Chr.2: 8987584 - 8989416 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127659.4 NP_565491.1 2 - 3 650 99
GenBank mRNA AY062799.1 - 2 - 3 561 99
AY081596.1 - 2 - 3 451 99

View Full Product Details