Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHT2;1
Entrez Gene ID
822265
Gene Name
phosphate transporter 2;1
Gene Aliases
AT3G26570, ORF02, phosphate transporter 2;1
Chromosome Location
Chr.3: 9756007 - 9758701 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113565.3 NP_189289.2 3 - 4 1284 72
NM_180302.2 NP_850633.1 2 - 3 1361 72
GenBank mRNA AF515591.1 - 2 - 3 952 72
AJ302645.1 - 2 - 3 1012 72
X97484.1 - - 1016 72

View Full Product Details