Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNM1
Entrez Gene ID
822280
Gene Name
DNA repair metallo-beta-lactamase family protein
Gene Aliases
AT3G26680, ATSNM1, SENSITIVE TO NITROGEN MUSTARD 1
Chromosome Location
Chr.3: 9801169 - 9804282 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338816.1 NP_001319649.1 7 - 8 1427 129
NM_001338817.1 NP_001319650.1 7 - 8 1427 129
NM_180304.2 NP_850635.1 7 - 8 1445 129
GenBank mRNA AK117422.1 - 7 - 8 1305 129
BT005000.1 - 7 - 8 1305 129

View Full Product Details