Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAB8
Entrez Gene ID
824529
Gene Name
RAB GTPase homolog 8
Gene Aliases
AT3G53610, ATRAB8, AtRABE1a, AtRab8B, GTPASE ATRAB8, RAB GTPase homolog 8
Chromosome Location
Chr.3: 19876232 - 19878792 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084809.2 NP_001078278.1 6 - 7 1006 127
NM_115221.4 NP_190929.1 7 - 8 883 127
NM_180365.3 NP_850696.1 7 - 8 556 127
GenBank mRNA AK228083.1 - 7 - 8 583 127
AK230120.1 - - 994 127
BT004779.1 - 6 - 7 478 127
U82434.1 - 7 - 8 498 127

View Full Product Details