Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BTR1L
Entrez Gene ID
830322
Gene Name
binding to TOMV RNA 1L (long form)
Gene Aliases
AT5G04430, BINDING TO TOMV RNA 1, BINDING TO TOMV RNA 1S (SHORT FORM), BTR1, BTR1S, T19N18.7, binding to TOMV RNA 1L (long form)
Chromosome Location
Chr.5: 1250248 - 1253953 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120525.4 NP_196063.1 2 - 3 376 59
NM_180433.3 NP_850764.1 2 - 3 376 59
GenBank mRNA AF375411.1 - 2 - 3 238 59
AY086553.1 - 2 - 3 256 59
BT000854.1 - 1 - 2 87 59

View Full Product Details