Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TOC34
Entrez Gene ID
830382
Gene Name
translocon at the outer envelope membrane of chloroplasts 34
Gene Aliases
ARABIDOPSIS THALIANA TRANSLOCON AT THE OUTER ENVELOPE MEMBRANE OF CHLOROPLASTS 34, AT5G05000, ATTOC34, MUG13.14, MUG13_14, OEP34, translocon at the outer envelope membrane of chloroplasts 34
Chromosome Location
Chr.5: 1474027 - 1476434 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120582.5 NP_196119.1 7 - 8 1000 86
NM_180437.2 NP_850768.1 7 - 8 882 86
NM_203003.3 NP_974732.1 6 - 7 1340 86
GenBank mRNA AK229425.1 - 7 - 8 943 86
BT044057.1 - 6 - 7 831 86

View Full Product Details