Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VAT1
Entrez Gene ID
831490
Gene Name
VALINE-TOLERANT 1
Gene Aliases
AT5G16290, MQK4.1, MQK4_1, VALINE-TOLERANT 1
Chromosome Location
Chr.5: 5333750 - 5337784 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121634.4 NP_197133.1 9 - 10 1255 65
NM_180498.3 NP_850829.1 9 - 10 1242 65
GenBank mRNA AK226906.1 - 9 - 10 1181 65
BT021989.1 - 9 - 10 1144 65

View Full Product Details