Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACA3
Entrez Gene ID
830296
Gene Name
alpha carbonic anhydrase 3
Gene Aliases
ALPHA CARBONIC ANHYDRASE 3, AT5G04180, ATACA3, F21E1.100, F21E1_100, alpha carbonic anhydrase 3
Chromosome Location
Chr.5: 1147648 - 1149284 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120500.3 NP_196038.1 3 - 4 539 73
GenBank mRNA AK221569.1 - 3 - 4 539 73
AY088872.1 - 3 - 4 535 73
DQ446916.1 - 3 - 4 492 73

View Full Product Details