Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SYTC
Entrez Gene ID
830301
Gene Name
Calcium-dependent lipid-binding (CaLB domain) family protein
Gene Aliases
AT5G04220, ATSYTC, F21E1.140, F21E1_140, NTMC2T1.3, NTMC2TYPE1.3, SYT3, synaptotagmin 3
Chromosome Location
Chr.5: 1155725 - 1158944 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342755.1 NP_001331162.1 10 - 11 1961 97
NM_120504.2 NP_568135.1 10 - 11 1845 97
NM_203000.2 NP_974729.1 10 - 11 1819 97
GenBank mRNA AB102952.1 - 10 - 11 1495 97
FM213367.1 - 10 - 11 1651 97
FM213368.1 - 10 - 11 1554 97

View Full Product Details