Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- KTF1
- Entrez Gene ID
-
830308
- Gene Name
- kow domain-containing transcription factor 1
- Gene Aliases
-
AT5G04290, SPT5-LIKE, SPT5L, T19N18.20, T19N18_20, kow domain-containing transcription factor 1
- Chromosome Location
-
Chr.5: 1195969 - 1202936 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_120511.1 |
NP_196049.1 |
15 - 16 |
2132 |
89 |
View Full Product Details