Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SIR
Entrez Gene ID
830336
Gene Name
sulfite reductase
Gene Aliases
AT5G04590, SULFITE REDUCTASE, T1E3.2, sulfite reductase
Chromosome Location
Chr.5: 1319074 - 1323364 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120541.4 NP_196079.1 1 - 2 521 64
GenBank mRNA AF325027.2 - 1 - 2 235 64

View Full Product Details