Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALA1
Entrez Gene ID
830375
Gene Name
aminophospholipid ATPase 1
Gene Aliases
AT5G04930, aminophospholipid ATPase 1
Chromosome Location
Chr.5: 1444638 - 1449879 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120575.3 NP_568146.1 5 - 6 3672 122

View Full Product Details