Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LAC12
Entrez Gene ID
830421
Gene Name
laccase 12
Gene Aliases
AT5G05390, K18I23.20, K18I23_20, laccase 12
Chromosome Location
Chr.5: 1594685 - 1597042 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120621.2 NP_196158.1 1 - 2 162 64

View Full Product Details